Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD2 Monoclonal Antibody (RPA-2.10), NovaFluor™ Blue 510, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | CD2 |
---|---|
Clone | RPA-2.10 |
Concentration | 0.2 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H072T03B01A
![]() |
Invitrogen™
H072T03B01A |
100 Tests |
Each for $430.00
|
|
|||||
H072T02B01A
![]() |
Invitrogen™
H072T02B01A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD2 Monoclonal antibody specifically detects CD2 in Mouse, Non-human Primate samples. It is validated for Flow CytometrySpecifications
CD2 | |
0.2 μg/Test | |
Monoclonal | |
Mouse | |
Mouse, Monkey | |
P08920 | |
12481 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
RPA-2.10 | |
Flow Cytometry | |
NovaFluor Blue 510 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Cd2; CD2 antigen; CD2 antigen (p50), sheep red blood cell receptor; CD2 molecule; CD2R; cluster of differentiation 2; Erythrocyte receptor; FLJ46032; LFA2; LFA-2; LFA-3 receptor; Ly37; Ly-37; lymphocyte antigen 37; OX34; OX-34; OX-34 antigen; OX-45 surface antigen homolog to human T lymphocyte CD2 antigen; Rosette receptor; SRBC; surface immunoglobulin superfamily molecule; T11; T-cell surface antigen CD2; T-cell surface antigen T11/Leu-5 | |
IgG1 κ | |
Affinity chromatography | |
CD2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title