Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD2 Mouse anti-Human,Baboon,Cynomolgus,Rhesus Ma, BB700, Clone: RPA-2.10, BD Horizon™
Mouse Monoclonal Antibody
$342.00
Specifications
Antigen | CD2 |
---|---|
Clone | RPA-2.10 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Blue 700 (BB700) |
Description
CD2 Monoclonal antibody specifically detects CD2 in Human,Baboon,Cynomolgus,Rhesus Ma samples. It is validated for Flow Cytometry.Specifications
CD2 | |
Flow Cytometry | |
Brilliant Blue 700 (BB700) | |
Mouse | |
Human, Baboon, Cynomolgus, Rhesus Macaque | |
Primary |
RPA-2.10 | |
Monoclonal | |
Purified | |
RUO | |
IgG1 κ | |
Refrigerate (2 to 8°C) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title