Learn More
Description
Specifications
Specifications
| Antigen | CD2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | RPA-2.10 |
| Conjugate | Brilliant Ultraviolet 496 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | LFA-2; LFA-3 receptor; SRBC; T-cell surface antigen T11/Leu-5; erythrocyte receptor |
| Host Species | Mouse |
| Immunogen | Human Phytohemagglutinin-treated Lymphoblasts |
| Quantity | 50 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.