Learn More
CD2 Mouse anti-Human, PE, Clone: RPA-2.10, BD
Mouse Monoclonal Antibody
$128.60 - $279.50
Specifications
Antigen | CD2 |
---|---|
Clone | RPA-2.10 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Description
The RPA-2.10 monoclonal antibody specifically binds to CD2. CD2 is a 50 kDa single-chain transmembrane glycoprotein, also known as LFA-2 or the receptor for sheep erythrocytes. CD2 belongs to the immunoglobulin superfamily of proteins along with its ligand LFA-3, CD58. It is present on about 80-90% of human peripheral blood lymphocytes, greater than 95% of thymocytes, all T lymphocytes that form E-rosettes and a subset of NK cells. CD2 plays a role in T-cell signaling and in lymphocyte adhesion.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
CD2 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified |
RPA-2.10 | |
Monoclonal | |
Mouse | |
Human | |
Human Phytohemagglutinin-treated Lymphoblasts | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.