Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD2 Mouse, anti-Human, R718, Clone: RPA-2.10, BD OptiBuild™

Catalog No. BDB752063 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752063 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752063 Supplier BD Biosciences Supplier No. 752063
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The RPA-2.10 monoclonal antibody specifically binds to CD2 which is also known as Lymphocyte-function antigen-2 (LFA-2), LFA-3 receptor, Erythrocyte receptor, Sheep red blood cell (SRBC) receptor, or T-cell surface antigen T11/Leu-5. CD2 is a 50 kDa type I transmembrane glycoprotein. CD2 belongs to the immunoglobulin superfamily of proteins along with its primary ligand, LFA-3 (CD58). It is expressed on the surface of ∼80-90% of human peripheral blood lymphocytes, greater than 95% of thymocytes, all T lymphocytes that form E-rosettes, and a subset of NK cells. CD2 functions as an adhesion receptor that binds to CD58 resulting in the activation of CD2-positive T cells and NK cells and in the regulation of their cytolytic activities.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD2
Applications Flow Cytometry
Classification Monoclonal
Clone RPA-2.10
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.