Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD2 Mouse, anti-Human, R718, Clone: S5.2, BD OptiBuild™

Catalog No. BDB752148 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752148 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752148 Supplier BD Biosciences Supplier No. 752148
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The S5.2 monoclonal antibody specifically recognizes the CD2 antigen, a 45 to 50 kDa type I transmembrane glycoprotein that forms the binding site for sheep erythrocytes. The CD2 antigen is present on approximately 75% of normal peripheral blood lymphocytes and 95% to 99% of thymocytes. It is also found on a subset of monocytes (approximately one-third) that might be precursors to dendritic cells. The CD2 antibody reacts with essentially all T lymphocytes and with a subset of NK lymphocytes. CD2 and CD58 have been shown to be coreceptors. The interaction of CD2 antigen and CD58 antigen facilitates antigen recognition by T lymphocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD2
Applications Flow Cytometry
Classification Monoclonal
Clone S5.2
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.