Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD2, Mouse Anti-Human, RB613, Clone: S5.2, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 759373
Description
CD2 Monoclonal antibody specifically detects CD2 in Human (Tested in Development) samples. It is validated for Flow Cytometry
Specifications
CD2 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg; | |
Primary | |
Human | |
IgG2a |
Flow Cytometry | |
S5.2 | |
RB613 | |
LFA-2; Lymphocyte function antigen-2; Rosette receptor; T11/Leu-5; Leu-5b | |
Alloantigen-activated T Cells | |
RUO | |
914 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction