Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD2 Mouse, anti-Rat, R718, Clone: OX-34, BD OptiBuild™

Catalog No. BDB752182 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752182 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752182 Supplier BD Biosciences Supplier No. 752182
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The OX-34 antibody reacts with CD2 (LFA-2), a member of the immunoglobulin superfamily. In the rat, CD2 is expressed on thymocytes, T lymphocytes in spleen and lymph node, dendritic epidermal T cells, splenic macrophages, and NK cells, but not on B cells, most intestinal intraepithelial lymphocytes, or peritoneal and liver macrophages. CD2 can associate with the T-cell receptor complex, and it may function in both intercellular adhesion and signal transduction. In the rat, CD48 and CD59 have been identified as ligands for CD2. OX-34 mAb binds to the extracellular portion of CD2, and it blocks the binding of CD2 to CD48. While OX-34 antibody does not activate T cells, it partially blocks activation by immobilized mAbs to CD3 (clone G4.18) and αβ T-cell receptor (clone R73), and it partially inhibits allogeneic mixed lymphocyte reactions. Moreover, in vivo administration of OX-34 antibody depletes peripheral T cells and prevents allograft rejection.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD2
Applications Flow Cytometry
Classification Monoclonal
Clone OX-34
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.