Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD2 Rabbit anti-Human, Clone: SP304, Abnova™

Catalog No. 89035363 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-363 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-363 Supplier Abnova Corporation Supplier No. MAB15149

Rabbit Monoclonal Antibody

Rabbit monoclonal antibody raised against partial recombinant human CD2.

  • High specificity
  • High reactivity

Specifications

Antigen CD2
Applications Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone SP304
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against partial recombinant human CD2.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene Alias SRBC/T11
Gene Symbols CD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 1-235 of human CD2.
Purification Method Protein A/G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 914
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.