Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD2, Rat anti-Human, Clone: YTH 655, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP2810420.025MG
Description
CD2 Monoclonal specifically detects CD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Cell depletion.Specifications
CD2 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry, Cell depletion | |
CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
Rat | |
Protein A purified | |
RUO | |
Primary | |
This antibody specifically recognizes human CD2, a cell adhesion molecule found on the surface of T cells and natural killer (NK) cells. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2b κ |
Flow Cytometry, Depletion Assay | |
YTH 655 | |
Unconjugated | |
PBS | |
CD2 | |
Recombinant monoclonal antibody to CD2. | |
0.025 mg | |
Adaptive Immunity, Apoptosis, Immunology | |
914 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction