Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD20 Ab-1, Mouse Monoclonal Antibody

Catalog No. MS340R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS340R7 7 mL Prediluted
MS340S0 100 μL Concentrated
MS340S1 500 μL Concentrated
MS340S 1 mL Concentrated
MS340RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS340R7 Supplier Epredia™ Supplier No. MS340R7
Add to Cart
Add to Cart

Obtain accurate and reproducible results in immunohistochemistry, immunofluorescence and flow cytometry applications.

CD20 is a non-Ig differentiation antigen of B cells and the expression of CD20 is restricted to normal and neoplastic B cells, being absent from all other leukocytes and tissues. It acts as calcium channel involved in B cell activation and cell cycle progression.

Host Species: Mouse

Clone: L26

Isotype: IgG2a/κ

Species Reactivity: Human. Does not react with cow, dog, pig and rat.

Epitope: Cytoplasmic domain

Immunogen: Human tonsil B cells

Molecular Weight: 30-33kDa

Positive Control: Tonsil

Cellular Localization: Membrane/cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry

Specifications

Antigen CD20 Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone L26
Conjugate Unconjugated
Host Species Mouse
Immunogen Human tonsil B cells
Quantity 7 mL
Regulatory Status IVD
Research Discipline B-Cell
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.