Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD20 Anti-Human BV421, Clone: Obinutuzumab297.rMAb (also known as Obinutuzumab N297A Biosimilar.rMAb), BD Horizon™
Mouse Monoclonal Antibody
$205.50 - $516.00
Specifications
| Antigen | CD20 |
|---|---|
| Clone | Obinutuzumab297.rMAb (also known as Obinutuzumab N297A Biosimilar.rMAb) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Violet 421 |
| Catalog Number | Mfr. No. | Quantity | Conjugate | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Conjugate | Price | Quantity & Availability | ||||
|
BDB570970
|
BD Biosciences
570970 |
100 Tests | Brilliant Violet 421 |
Each for $516.00
|
|
||||
|
BDB571041
|
BD Biosciences
571041 |
25 Tests | Brilliant Violet 421 |
Each for $205.50
|
|
||||
Description
CD20 Monoclonal antibody specifically detects CD20 in Human (QC Testing) samples. It is validated for Flow CytometrySpecifications
| CD20 | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| Biosimilar | |
| Human | |
| B1; Bp35; LEU-16; MS4A1 | |
| Human CD20 | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Obinutuzumab297.rMAb (also known as Obinutuzumab N297A Biosimilar.rMAb) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| 931 | |
| IgG1 κ | |
| The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title