Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD20 Antibody (10F381 (rituximab)) - Chimeric - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. p-7106014 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25264625 0.025 mg
NBP25264602 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25264625 Supplier Novus Biologicals Supplier No. NBP2526460.025MG
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD20 Monoclonal specifically detects CD20 in Human, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen CD20
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 10F381 (rituximab)
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000, CyTOF-ready
Formulation PBS with 0.02% Proclin 300. with 0.02% Proclin 300
Gene Alias B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2
Gene Symbols MS4A1
Host Species Rabbit
Immunogen Human lymphoblastoid cell line SB.
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers
Primary or Secondary Primary
Gene ID (Entrez) 931
Test Specificity CD20 (Human B-lymphocyte-restricted differentiation antigen, Bp35), is a hydrophobic transmembrane protein with a molecular weight of approximately 35 kDa located on pre-B and mature B lymphocytes, and most B-cell non-Hodgkin's lymphomas, but is not found on stem cells, pro-B cells, normal plasma cells or other normal tissues .
Target Species Human
Content And Storage Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.