Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    CD20 Antibody (396444), mFluor Violet 610 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4225MFV610
Description
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
| CD20 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers | |
| 931 | |
| Store at 4C in the dark. | 
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 396444 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 | |
| NS0 mouse myeloma cell line transfected with human CD20, Met1-Pro297, Accession # P11836 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            