Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD20 Antibody (AISB12), PE/Cy7, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143435PECY7
Description
CD20 Monoclonal specifically detects CD20 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
CD20 | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
MS4A1 | |
The immunogen for this antibody was CD20. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2a |
Flow Cytometry | |
AISB12 | |
Flow Cytometry | |
B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 | |
Rat | |
Protein A or G purified | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers | |
931.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction