Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD20 Antibody (10F381 (rituximab)), DyLight 350, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP275897UV
Description
CD20 Monoclonal specifically detects CD20 in Human, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Flow Cytometry.Specifications
CD20 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MS4A1 | |
Human lymphoblastoid cell line SB. | |
0.1 mL | |
Primary | |
CD20 (Human B-lymphocyte-restricted differentiation antigen, Bp35), is a hydrophobic transmembrane protein with a molecular weight of approximately 35 kDa located on pre-B and mature B lymphocytes, and most B-cell non-Hodgkin′s lymphomas, but is not found on stem cells, pro-B cells, normal plasma cells or other normal tissues . | |
Store at 4°C in the dark. |
Flow Cytometry | |
10F381 (rituximab) | |
Flow Cytometry | |
B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 | |
Human | |
Protein A purified | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers | |
931.0 | |
Human, Cynomolgus Monkey, Rhesus Macaque | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction