Learn More
CD20 Monoclonal Antibody (C273), Abbomax
Mouse Monoclonal Antibody
Supplier: Abbomax Inc 50011214
Description
Positive control: Tonsil Cellular location: Cell membrane.
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CD20 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with proprietary stabilizer and 0.01% sodium azide | |
| P11836 | |
| Mouse | |
| Protein A | |
| RUO | |
| 931 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| C273 | |
| Unconjugated | |
| Ms4a1 | |
| AA960661; B1; B-cell differentiation antigen Ly-44; B-lymphocyte antigen CD20; B-lymphocyte cell-surface antigen B1; B-lymphocyte surface antigen B1; Bp35; Cd20; CD20 antigen; CD20 cell surface protein; CD20 receptor; CVID5; EGK_06167; LEU-16; Leukocyte surface antigen Leu-16; Ly-44; Lymphocyte antigen 44; membrane spanning 4-domains A1; membrane-spanning 4-domains subfamily A member 1; membrane-spanning 4-domains, subfamily A, member 1; membrane-spanning 4-domains, subfamily A, member 2; MGC3969; MS4A1; MS4A2; S7 | |
| A synthetic peptide corresponding to C-terminus of human CD20 protein | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.