Learn More
Invitrogen™ CD20 Monoclonal Antibody (SP32)
Rabbit Monoclonal Antibody
$745.00 - $1137.00
Specifications
Antigen | CD20 |
---|---|
Clone | SP32 |
Concentration | Conc. not determined |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA139416
|
Invitrogen™
MA139416 |
1 mL |
Each for $1,137.00
|
|
|||||
PIMA516334
|
Invitrogen™
MA516334 |
500 μL |
Each for $745.00
|
|
|||||
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is tonsil or B cell lymphoma.
CD20 is a non-glycosylated surface phosphoprotein that has a molecular weight range of 33-37 kDa depending on the degree of phosphorylation. CD20 is expressed on mature and most malignant B cells, in a subpopulation of T lymphocytes and follicular dendritic cells. CD20 expression on B cells is synchronous with the expression of surface IgM and it regulates transmembrane calcium conductance, cell cycle progression and B-cell proliferation. CD20 is also associated with lipid rafts, but the intensity of this association depends on extracellular triggering, employing CD20 conformational change, and/or BCR (B cell antigen receptor) aggregation. After the receptor ligation, BCR and CD20 colocalize and then rapidly dissociate before BCR endocytosis, whereas CD20 remains at the cell surface. CD20 serves as a useful target for antibody-mediated therapeutic depletion of B cells, as it is expressed at high levels on most B-cell malignancies, but does not become internalized or shed from the plasma membrane following monoclonal antibody treatment. Diseases associated with CD20 dysfunction include Ms4a1-related common variable immune deficiency.Specifications
CD20 | |
Conc. not determined | |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
P11836, P19437 | |
12482, 931 | |
Synthetic peptide from C-terminus of human CD20 protein. | |
Antibody |
SP32 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Monoclonal | |
Liquid | |
IgG | |
Human, Mouse | |
Ms4a1 | |
AA960661; APY; ATOPY; B1; B-cell differentiation antigen Ly-44; B-lymphocyte antigen CD20; B-lymphocyte cell-surface antigen B1; B-lymphocyte surface antigen B1; Bp35; Cd20; CD20 antigen; CD20 cell surface protein; CD20 receptor; CVID5; EGK_06167; Fc epsilon receptor I beta chain; Fc Fragment of IgE high affinity I receptor for beta polypeptide; FCER1B; High affinity immunoglobulin epsilon receptor subunit beta; IgE Fc receptor subunit beta; IGEL; IGER; IGHER; LEU16; LEU-16; Leukocyte surface antigen Leu-16; Ly44; Ly-44; Lymphocyte antigen 44; membrane spanning 4-domains A1; membrane-spanning 4-domains subfamily A member 1; membrane-spanning 4-domains, subfamily A, member 1; membrane-spanning 4-domains, subfamily A, member 2; MGC3969; MS4A1; MS4A2; S7 | |
Ms4a1 | |
Primary | |
Protein A/G |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.