Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD20 Mouse anti-Baboon, Human, Rhesus, , Brilliant Ultraviolet 737, Clone: 2H7, BD Horizon™
Mouse Monoclonal Antibody
$248.00 - $472.00
Specifications
Antigen | CD20 |
---|---|
Clone | 2H7 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Ultraviolet 737 |
Description
CD20 Monoclonal antibody specifically detects CD20 in Baboon,Human,Rhesus, samples. It is validated for Flow Cytometry.Specifications
CD20 | |
Flow Cytometry | |
Brilliant Ultraviolet 737 | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2b κ | |
Affinity Purified |
2H7 | |
Monoclonal | |
Mouse | |
Baboon, Human, Rhesus Monkey | |
Human 6.16c1.3 B cell line | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title