Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD20 Mouse anti-Human, Feline, AF 700, Clone: B-Ly1, Southern Biotech™
SDP

Catalog No. OB935027 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB935027 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB935027 Supplier Southern Biotechnology Associates Supplier No. 935027
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SouthernBiotech anti-human primary antibodies are available in a variety of conjugated formats including Alexa Fluor and Pacific Blue.

Specifications

Antigen CD20
Applications Flow Cytometry
Classification Monoclonal
Clone B-Ly1
Concentration Lot specific
Conjugate Alexa Fluor 700
Formulation Supplied in PBS/NaN3
Host Species Mouse
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Feline, Human
Content And Storage Store at 2-8°C; Protect from light
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.