Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD20 Mouse anti-Human, Baboon, Cynomolgus, Rhesus, BUV805, Clone: 2H7, BD Horizon™
Mouse Monoclonal Antibody
$249.00 - $472.00
Specifications
| Antigen | CD20 |
|---|---|
| Clone | 2H7 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Ultraviolet 805 |
Description
CD20 Monoclonal antibody specifically detects CD20 in Human,Baboon,Cynomolgus,Rhesus samples. It is validated for Flow Cytometry.Specifications
| CD20 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 805 | |
| RUO | |
| IgG2b κ | |
| Refrigerate (2 to 8°C) |
| 2H7 | |
| Monoclonal | |
| Mouse | |
| Human, Baboon, Cynomolgus, Rhesus Monkey | |
| Primary | |
| CD20 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title