Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD20 Mouse anti-Human, Feline, R-PE, Clone: B-Ly1, Southern Biotech™

Mouse Monoclonal Antibody
$135.70 - $310.00
Specifications
| Antigen | CD20 |
|---|---|
| Clone | B-Ly1 |
| Concentration | Lot specific |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
SouthernBiotech anti-human primary antibodies are available in a variety of conjugated formats including Alexa Fluor and Pacific Blue.
Specifications
| CD20 | |
| Lot specific | |
| Monoclonal | |
| Mouse | |
| Feline, Human | |
| IgG1 | |
| Store at 2-8°C; Protect from light; Do not freeze |
| B-Ly1 | |
| Flow Cytometry | |
| R-PE | |
| RUO | |
| Supplied in PBS/NaN3 and a stabilizing agent | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title