Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD20, Mouse Anti-Human, RB613, Clone: 2H7, BD Horizon™
Mouse Monoclonal Antibody
$166.00 - $370.00
Specifications
Antigen | CD20 |
---|---|
Clone | 2H7 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB613 |
Description
CD20 Monoclonal antibody specifically detects CD20 in Human (QC Testing), Rhesus,Cynomolgus,Baboon (Tested in Development) samples. It is validated for Flow CytometrySpecifications
CD20 | |
Flow Cytometry | |
RB613 | |
Mouse | |
Human, Rhesus Monkey, Cynomolgus, Baboon | |
MS4A1; B1; Bp35; LEU-16; S7 | |
Human 6.16c1.3 B cell line | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
2H7 | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
931 | |
IgG2b κ | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title