Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD20, Mouse Anti-Human, RY610, Clone: 2H7, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 571163
Description
CD20 Monoclonal antibody specifically detects CD20 in Human (QC Testing), Rhesus,Cynomolgus,Baboon (Tested in Development) samples. It is validated for Flow Cytometry
Specifications
CD20 | |
Monoclonal | |
RY610 | |
MS4A1; B1; Bp35; LEU-16; S7 | |
Human 6.16c1.3 B cell line | |
25 Tests | |
Primary | |
Human, Rhesus Monkey, Cynomolgus, Baboon | |
Purified |
Flow Cytometry | |
2H7 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. | |
RUO | |
931 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction