Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD20 (Obinutuzumab) Recombinant Human Monoclonal Antibody (GA101), Functional Grade, Leinco Technologies™
SDP

Catalog No. 502544116
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4116 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4116 Supplier Leinco Technologies Inc Supplier No. LT906100MG
Add to Cart
Add to Cart

Human Recombinant Monoclonal Antibody

Specificity: Obinutuzumab (GA101) activity is directed against human CD20. Endotoxin Level: <1.0 EU/mg as determined by the LAL method. Purity: ≥98% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: Functional grade biosimilar antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.

CD20 (Obinutuzumab) Recombinant Monoclonal antibody specifically detects CD20 (Obinutuzumab) in Cynomolgus Monkey, Human, Rhesus Monkey samples. It is validated for ELISA, Flow Cytometry, Functional Assay, Western Blot

Specifications

Antigen CD20 (Obinutuzumab)
Applications ELISA, Flow Cytometry, Functional Assay, Western Blot
Classification Recombinant Monoclonal
Clone GA101
Concentration ≥5.0 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2-7.4
Gene Alias afutuzumab; Gazyva; Gazyvaro; RG7159; RO5072759
Host Species Human
Immunogen Human lymphoblastoid cell line SB
Purification Method Protein A/G
Quantity 100 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Cynomolgus Monkey, Human, Rhesus Monkey
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.