Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ CD20 Rabbit Monoclonal Antibody supernatant

Catalog No. RM2115R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
100 μL
12 mL
501 mL
7 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RM2115R7 7 mL
RM2115S0 100 μL
RM2115R1 1 mL
RM2115RQ 12 mL
RM2115S 501 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RM2115R7 Supplier Epredia™ Supplier No. RM2115R7

CD20, Rabbit Monoclonal Ab is part of a comprehensive ab portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

CD20 is a non-Ig differentiation antigen of B cells and the expression of CD20 is restricted to normal and neoplastic B cells, being absent from all other leukocytes and tissues. It acts as calcium channel involved in B cell activation and cell cycle progression.

Host Species: Rabbit

Clone: EP459Y

Isotype: Rabbit IgG

Species Reactivity: Human. Others not tested.

Immunogen: A synthetic peptide corresponding the residues in the C-term of human CD20

Molecular Weight: 35kDa

Positive Control: Tonsil

Cellular Localization: Cell Membrane/cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EP459Y
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide corresponding the residues in the C-term of human CD20
Quantity 7 mL
Regulatory Status IVD
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.