Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD20 Rabbit Polyclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry, western blotting, and immunoprecipitation experiments with Epredia™ CD20, Rabbit Polyclonal Antibody.

$16590.00

Specifications

Antigen CD20
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 4
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
RB9013P
View Documents
Epredia
RB9013P
Concentrated 1 mL
Each for $16,590.00
Only null left
Add to Cart
 
RB9013P0
View Documents
Epredia
RB9013P0
Concentrated 100 μL N/A N/A N/A
RB9013P1
View Documents
Epredia
RB9013P1
Concentrated 500 μL N/A N/A N/A
RB9013R7
View Documents
Epredia
RB9013R7
Prediluted 7 mL N/A N/A N/A
Description

Description

CD20 is a 33kDa leukocyte surface antigen consisting of four transmembrane regions and cytoplasmic N- and C-termini. CD20 is expressed primarily on B cells but has also been detected on both normal and neoplastic T cells. Activation of CD20 significantly increases DNA synthesis and is thought to involve basic helix-loop-helix leucine zipper transcription factors.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: C-terminus

Immunogen: Synthetic peptide derived from C-terminus of human CD20 protein

Molecular Weight: 33-37kDa

Positive Control: Tonsil

Cellular Localization: Membrane/cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation
Specifications

Specifications

CD20
Polyclonal
Rabbit
B-Cell
Synthetic peptide derived from C-terminus of human CD20 protein
Primary
Immunoprecipitation, Western Blot
Unconjugated
IVD
Human
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.