Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD20 Rat anti-Mouse, Alexa Fluor™ 488, Clone: GOT214A, BD Phrmingen™
Rat Monoclonal Antibody
$164.00 - $367.50
Specifications
Antigen | CD20 |
---|---|
Clone | GOT214A |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD20 Monoclonal antibody specifically detects CD20 in Mouse samples. It is validated for Flow cytometrySpecifications
CD20 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
RBL-1-mCD20 transfected cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
GOT214A | |
Flow Cytometry | |
Alexa Fluor 488 | |
Rat | |
Mouse | |
Ly-44; Ms4a1; Ms4a2 | |
IgG2a κ | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title