Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD20 Rituximab Biosimilar anti-Human, Unconjugated, Clone: Rituximab.rMAb (also known as Rituximab Biosimilar), BD Phrmingen™
Biosimilar Monoclonal Antibody
Supplier: BD Biosciences 569496
Description
CD20 Rituximab Biosimilar Monoclonal antibody specifically detects CD20 Rituximab Biosimilar in Human samples. It is validated for Flow cytometry|Cytotoxicity
Specifications
| CD20 Rituximab Biosimilar | |
| Monoclonal | |
| 1.0 mg/mL | |
| No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2 μm sterile filtered. Endotoxin level is ≤0.1 EU/μg (≤0.01 ng/μg) of protein as determined by the LAL assay. | |
| Human CD20 | |
| 0.5 mg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Cytotoxicity Assay | |
| Rituximab.rMAb (also known as Rituximab Biosimilar) | |
| Unconjugated | |
| B1; Bp35; LEU-16; MS4A1 | |
| Affinity Chromatography | |
| RUO | |
| 931 | |
| Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. This preparation contains no preservatives, thus it should be handled under aseptic conditions. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction