Learn More
Invitrogen™ CD200 Monoclonal Antibody (325516), PE

Description
In direct ELISAs, no cross-reactivity with recombinant mouse CD200 is observed.
Specifications
Specifications
| Antigen | CD200 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 325516 |
| Conjugate | PE |
| Formulation | Buffered saline solution with ≤ 0.5% BSA and 0.09% sodium azide |
| Gene | Cd200 |
| Gene Accession No. | P41217 |
| Gene Alias | antigen identified by monoclonal antibody MRC OX-2; CD200; Cd200 antigen; Cd200 molecule; cell surface protein (thymocyte antigen identified by monoclonal antibody MRC-OX2); Cspmo2; hypothetical protein LOC534910; MOX1; MOX2; MRC; MRC OX-2 antigen; MRCOX2; My033; OX2; OX-2; OX-2 membrane glycoprotein |
| Gene Symbols | Cd200 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.