Learn More
Invitrogen™ CD200 Monoclonal Antibody (MRC OX104), FITC

Description
OX-104 recognizes the human CD200 cell surface antigen, also known as OX2.CD200 is expressed by a subset of B lymphocytes, some endothelial cells and by neurons. Studies have suggested that the CD200-CD200 ligand system is of importance in the control of macrophage and granulocyte activation.
Specifications
Specifications
| Antigen | CD200 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MRC OX104 |
| Concentration | 0.1 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 1% BSA and 0.09% sodium azide |
| Gene | Cd200 |
| Gene Accession No. | P41217 |
| Gene Alias | antigen identified by monoclonal antibody MRC OX-2; CD200; Cd200 antigen; Cd200 molecule; cell surface protein (thymocyte antigen identified by monoclonal antibody MRC-OX2); Cspmo2; hypothetical protein LOC534910; MOX1; MOX2; MRC; MRC OX-2 antigen; MRCOX2; My033; OX2; OX-2; OX-2 membrane glycoprotein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.