Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD200 Monoclonal Antibody (OX-2), Biotin

Catalog No. PIMA517564
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517564 500 μg
PIMA517563 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA517564 Supplier Invitrogen™ Supplier No. MA517564
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD200 Monoclonal Antibody for Flow

CD200 is a type-1 membrane glycoprotein in the immunoglobulin superfamily, featuring two immunoglobulin domains. It is widely expressed on various cells, including B and T cells, keratinocytes, nerve cells, follicular dendritic cells, ovarian cells, and trophoblasts. CD200 interacts with CD200R on myeloid cells, inhibiting macrophage activation, reducing IL-6 production, and attenuating lymphocyte activation. It also prevents mast cell degranulation and decreases TNF-alpha and IL-13 secretion, playing a role in immune regulation and maternal tolerance. Homologues of CD200 are found in herpesviruses and poxviruses, indicating a conserved immune modulation function. CD200 has multiple alternatively spliced transcript variants, contributing to its functional diversity. Diseases linked to CD200 dysfunction include Mantle Cell Lymphoma and Primary Hypertrophic Osteoarthropathy, highlighting its importance in health and disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD200
Applications Flow Cytometry
Classification Monoclonal
Clone OX-2
Concentration 0.1 mg/mL
Conjugate Biotin
Formulation PBS with 4-5mg/mL BSA and 0.02% sodium azide
Gene Cd200
Gene Accession No. P04218
Gene Alias antigen identified by monoclonal antibody MRC OX-2; CD200; Cd200 antigen; Cd200 molecule; cell surface protein (thymocyte antigen identified by monoclonal antibody MRC-OX2); Cspmo2; hypothetical protein LOC534910; MOX1; MOX2; MRC; MRC OX-2 antigen; MRCOX2; My033; OX2; OX-2; OX-2 membrane glycoprotein
Gene Symbols Cd200
Host Species Mouse
Immunogen Rat thymocyte membrane glycoproteins were fused with mouse NS-1 myeloma cells.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24560
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.