Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD200 Monoclonal Antibody (OX104), NovaFluor™ Yellow 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | CD200 |
---|---|
Clone | OX104 |
Concentration | 0.6 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H038T03Y06A
![]() |
LIFE TECHNOLOGIES
H038T03Y06A |
100 Tests |
Each for $430.00
|
|
|||||
H038T02Y06A
![]() |
LIFE TECHNOLOGIES
H038T02Y06A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD200 Monoclonal antibody specifically detects CD200 in Human samples. It is validated for Flow CytometrySpecifications
CD200 | |
0.6 μg/Test | |
Monoclonal | |
Mouse | |
Human | |
P41217 | |
4345 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
OX104 | |
Flow Cytometry | |
NovaFluor Yellow 700 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
antigen identified by monoclonal antibody MRC OX-2; CD200; Cd200 antigen; Cd200 molecule; cell surface protein (thymocyte antigen identified by monoclonal antibody MRC-OX2); Cspmo2; hypothetical protein LOC534910; MOX1; MOX2; MRC; MRC OX-2 antigen; MRCOX2; My033; OX2; OX-2; OX-2 membrane glycoprotein | |
IgG1 κ | |
Affinity Chromatography | |
Cd200 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title