Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD200 Mouse anti-Human , APC, Clone: MRC OX-104, BD Biosciences™
Mouse Monoclonal Antibody
$144.00 - $345.50
Specifications
Antigen | CD200 |
---|---|
Clone | MRC OX-104 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC |
Description
CD200 Monoclonal antibody specifically detects CD200 in Human samples. It is validated for Flow CytometrySpecifications
CD200 | |
Flow Cytometry | |
APC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human CD200 Recombinant Protein | |
Primary |
MRC OX-104 | |
Monoclonal | |
Mouse | |
Human | |
OX-2; MOX1; MOX2 ; My033 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to APC under optimum conditions, and unconjugated antibody and free APC were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title