Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200 Mouse anti-Human, Brilliant Violet 650, Clone: MRC OX-104, BD Optibuild™

Catalog No. BDB743085 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743085 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743085 Supplier BD Biosciences Supplier No. 743085
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MRC OX-104 monoclonal antibody specifically binds to CD200. CD200 is a 40-45 kDa type 1 transmembrane glycoprotein and is also referred to as OX2. CD200 is a member of the immunoglobulin superfamily of proteins. It contains two Ig domains, a single transmembrane region and a short cytoplasmic tail. CD200 is expressed on some dendritic cell subsets, and resting and activated T- and B-cells, but not on NK cells, monocytes, granulocytes, or platelets. It has been found on a subset of CD34+ progenitor cells. Interaction of CD200 with its receptor on macrophages induces a downregulation of macrophage activity in a variety of tissues, suggesting a regulatory function.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD200
Classification Monoclonal
Clone MRC OX-104
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias OX-2, MOX1, MOX2, My033
Host Species Mouse
Immunogen Human CD200 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.