Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200 Mouse anti-Human, Clone:6E8B11, Abnova™

Catalog No. 89333186 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-186 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-186 Supplier Abnova Corporation Supplier No. MAB16609
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human CD200.

The protein encoded by this gene is a type-1 membrane glycoprotein, which contains two immunoglobulin domains, and thus belongs to the immunoglobulin superfamily. Studies of the related genes in mouse and rat suggest that this gene may regulate myeloid cell activity and delivers an inhibitory signal for the macrophage lineage in diverse tissues. Multiple alternatively spliced transcript variants that encode different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Antigen CD200
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6E8B11
Conjugate Unconjugated
Description CD200 molecule
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene CD200
Gene Alias MOX1, MOX2, MRC, OX-2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 56-257 of human CD200 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 4345
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.