Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD200/OX2 Antibody (325516), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB27241L
Description
CD200/OX2 Monoclonal antibody specifically detects CD200/OX2 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
CD200/OX2 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
325516 | |
Flow Cytometry, CyTOF-ready | |
antigen identified by monoclonal MRC OX-2, CD200 antigenMOX1, CD200 molecule, MOX2MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein | |
Mouse myeloma cell line NS0-derived recombinant human CD200, Gln31-Gly232, Accession # P41217.3 | |
0.1 mL | |
Immunology | |
4345 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction