Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200 Receptor Mouse anti-Human, Brilliant Violet 421, Clone: OX-108 (also known as OX108), BD

Catalog No. BDB566344 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566344 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566344 Supplier BD Biosciences Supplier No. 566344
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The OX-108 monoclonal antibody specifically binds to CD200R which is also known as OX2 receptor 1 (OX2R) or MOX2 receptor (MOX2R). CD200R is a type I transmembrane glycoprotein that is encoded by CD200R1 (CD200 receptor 1) and belongs to the C2-set domain containing gene family within the Ig superfamily. CD200R functions as an inhibitory receptor when bound by its ligand, CD200. CD200R is expressed by hematopoietic cells including monocytes, macrophages, dendritic cells, Langerhans cells, neutrophils, basophils, most T cells and mast cells. CD200R signaling may regulate the functions of various types of leucocytes.

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD200 Receptor
Classification Monoclonal
Clone OX-108 (also known as OX108)
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias CD200R1, CD200 receptor 1, hCD200R, CRTR2, HCRTR2, MOX2R, OX2R
Host Species Mouse
Immunogen Human CD200R Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.