Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200R Rat anti-Mouse, Alexa Fluor 647, Clone: OX-110 (also known as OX110), BD

Catalog No. BDB566345 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566345 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566345 Supplier BD Biosciences Supplier No. 566345
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The OX-110 monoclonal antibody specifically binds to CD200R which is also known as OX2 receptor 1 (Ox2r) or MOX2 receptor (Mox2r). CD200R is a type I transmembrane glycoprotein that is encoded by Cd200r1 (CD200 receptor 1) and belongs to the C2-set domain containing gene family within the Ig superfamily. CD200R is expressed by hematopoietic cells including monocytes, granulocytes, and T cells. CD200R functions as an inhibitory receptor when bound by its ligand, CD200. CD200R signaling can inhibit leucocyte functions such as the production of cytokines.

Alexa Fluor™ 647 conjugates are highly photostable and remain fluorescent over a broad pH range. The excitation and emission maxima are nearly identical to those of APC. However, APC tends to be brighter while Alexa Fluor™ 647 is more optimal for intracellular applications. This fluorochrome exhibits uncommon photostability, making it an ideal choice for use in fluorescence microscopy. Due to nearly identical excitation and emission properties but different spillover characteristics, APC and Alexa Fluor™ 647 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD200R
Classification Monoclonal
Clone OX-110 (also known as OX110)
Concentration 0.2mg/mL
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Cd200r1, CD200 receptor 1, Orexin receptor 2, Ox2r, Mox2r, MO2R1
Host Species Rat
Immunogen Mouse CD200R Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.