Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD200R Recombinant Rabbit Monoclonal Antibody (019)
GREENER_CHOICE

Catalog No. PIMA530237
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530237 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530237 Supplier Invitrogen™ Supplier No. MA530237
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse CD200R1.

CD200R (OX2R) is a receptor for the OX-2 membrane glycoprotein, and both the receptor and substrate are cell surface glycoproteins containing two immunoglobulin-like domains. CD200R is restricted to the surfaces of myeloid lineage cells and the receptor-substrate interaction may function as a myeloid downregulatory signal. In mouse, CD200R is expressed primarily by peripheral blood monocytes and neutophils but also by other leucocytes including T-lymphocytes and mast cells. Further mouse studies of a gene related to CD200R suggests that the receptor may control myeloid function in a tissue-specific manner. Alternative splicing of CD200R results in multiple transcript variants. Diseases associated with CD200R dysfunction include primary hypertrophic osteoarthropathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD200R
Applications ELISA
Classification Recombinant Monoclonal
Clone 019
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CD200R1
Gene Accession No. Q9ES57
Gene Alias antigen identified by monoclonal antibody MRC OX-2 receptor; CD200 cell surface glycoprotein receptor; CD200 R1; CD200 receptor 1; CD200R; Cd200r1; Cell surface glycoprotein CD200 receptor 1; cell surface glycoprotein OX2 receptor 1; cell surface glycoprotein receptor CD200; CRTR2; HCRTR2; MOX2 receptor; MOX2R; Orexin receptor 2; OX102 antigen; OX2 receptor; OX2R; UNQ2522/PRO6015
Gene Symbols CD200R1
Host Species Rabbit
Immunogen Recombinant Mouse CD200R1 protein (Met1-Pro238).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57781
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.