Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD200R (soluble) Monoclonal Antibody (OX110), FITC
GREENER_CHOICE

Catalog No. PIMA517984
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517984 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517984 Supplier Invitrogen™ Supplier No. MA517984
Add to Cart
Add to Cart

Rat Monoclonal Antibody

MA5-17984 targets CD200R/OX-2R in FACS applications and shows reactivity with Mouse samples. The MA5-17984 immunogen is soluble recombinant mouse CD200R.

CD200R (OX2R) is a receptor for the OX-2 membrane glycoprotein, and both the receptor and substrate are cell surface glycoproteins containing two immunoglobulin-like domains. CD200R is restricted to the surfaces of myeloid lineage cells and the receptor-substrate interaction may function as a myeloid downregulatory signal. In mouse, CD200R is expressed primarily by peripheral blood monocytes and neutophils but also by other leucocytes including T-lymphocytes and mast cells. Further mouse studies of a gene related to CD200R suggests that the receptor may control myeloid function in a tissue-specific manner. Alternative splicing of CD200R results in multiple transcript variants. Diseases associated with CD200R dysfunction include primary hypertrophic osteoarthropathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD200R (soluble)
Applications Flow Cytometry
Classification Monoclonal
Clone OX110
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 4-5mg/mL BSA and 0.02% sodium azide
Gene CD200R1
Gene Accession No. Q9ES57
Gene Alias antigen identified by monoclonal antibody MRC OX-2 receptor; CD200 cell surface glycoprotein receptor; CD200 R1; CD200 receptor 1; CD200R; Cd200r1; Cell surface glycoprotein CD200 receptor 1; cell surface glycoprotein OX2 receptor 1; cell surface glycoprotein receptor CD200; CRTR2; HCRTR2; MOX2 receptor; MOX2R; Orexin receptor 2; OX102 antigen; OX2 receptor; OX2R; UNQ2522/PRO6015
Gene Symbols CD200R1
Host Species Rat
Immunogen Soluble recombinant mouse CD200R.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57781
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.