Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200R1, anti-Human, Clone: OX108, Abnova™

Catalog No. 89032353 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-032-353 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-353 Supplier Abnova Corporation Supplier No. RAB00010
Add to Cart
Add to Cart

Monoclonal Antibody

Specifications

Antigen CD200R1
Applications Flow Cytometry
Classification Monoclonal
Clone OX108
Conjugate Unconjugated
Dilution Flow Cytometry (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.02% Proclin 300).
Gene CD200 receptor 1
Gene Alias CD200R/HCRTR2/MOX2R/OX2R
Gene Symbols CD200R1
Immunogen Original antibody is raised against fusion protein hCD200RCD4d3+4.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131450
Test Specificity This antibody recognizes human CD200R1.
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.