Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD200R1 Antibody (118), DyLight 488 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289845G
Description
CD200R1 Monoclonal antibody specifically detects CD200R1 in Human samples. It is validated for ELISASpecifications
CD200R1 | |
Monoclonal | |
DyLight 488 | |
CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, OX2RCD200 cell surface glycoprotein receptor | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD200R1 (AAI43394.1; Met1-Leu266). | |
0.1 mL | |
Cancer, Neuronal Cell Markers, Neuroscience | |
131450 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
118 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction