Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD200R1 Antibody (OX-110), Alexa Fluor™ 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10063562AF405
Description
CD200R1 Monoclonal specifically detects CD200R1 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD200R1 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD200R1 | |
Fusion protein mCD200RCD4d3+4 | |
0.1 mL | |
Cancer, GPCR, Neuronal Cell Markers, Neuroscience | |
131450 | |
Mouse | |
Purified |
Flow Cytometry | |
OX-110 | |
Flow Cytometry | |
CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, OX2RCD200 cell surface glycoprotein receptor | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
NB100-63562 recognizes mouse CD200R, a cell surface glycoprotein (also known as OX2R), In mouse CD200R is expressed primarily by peripheral blood monocytes and neutrophils but also by other leucocytes including T-lymphocytes and mast cells (1). Studies suggest that CD200-CD200R interaction may be involved in the control of myeloid cellular function. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction