Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD200R1 Antibody (OX108), Alexa Fluor™ 532 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP252642AF532
Description
CD200R1 Monoclonal specifically detects CD200R1 in Human samples. It is validated for Flow Cytometry.Specifications
CD200R1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
CD200R1 | |
Recombinant protein - hCD200RCD4d3+4. | |
Primary | |
Recognizes human CD200R, a receptor for the CD200/OX2. | |
Store at 4°C in the dark. |
Flow Cytometry | |
OX108 | |
Flow Cytometry | |
CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, OX2RCD200 cell surface glycoprotein receptor | |
Rabbit | |
0.1 mL | |
131450 | |
Human | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction