Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200R1 Antibody (OX108) - Chimeric - Azide and BSA Free, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$235.84 - $615.90

Specifications

Antigen CD200 R1
Clone OX108
Dilution Flow Cytometry 1:10 - 1:1000, CyTOF-ready
Applications Flow Cytometry, CyTOF
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25264225
SDP
View Documents
Novus Biologicals
NBP2526420.025MG
0.025 mg
Each for $235.84
Only null left
Add to Cart
 
NBP25264202
SDP
View Documents
Novus Biologicals
NBP2526420.2MG
0.2 mg
Each for $615.90
Only null left
Add to Cart
 
Description

Description

CD200R1 Monoclonal specifically detects CD200R1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications

Specifications

CD200 R1
Flow Cytometry 1:10 - 1:1000, CyTOF-ready
Monoclonal
Purified
RUO
Human
CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, OX2RCD200 cell surface glycoprotein receptor
CD200R1
IgG
Protein A or G purified
Recognizes human CD200R, a receptor for the CD200/OX2.
OX108
Flow Cytometry, CyTOF
Unconjugated
Rabbit
Cancer, GPCR, Neuronal Cell Markers, Neuroscience
PBS with 0.02% Proclin 300. with 0.02% Proclin 300
131450
Recombinant protein - hCD200RCD4d3+4.
Primary
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.