Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD200R1 Rabbit anti-Mouse, Clone: 181, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
| Antigen | CD200R1 |
|---|---|
| Clone | 181 |
| Dilution | Flow Cytometry 1:25-1:100 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
CD200R1 Monoclonal specifically detects CD200R1 in Mouse samples. It is validated for Flow Cytometry.Specifications
| CD200R1 | |
| Flow Cytometry 1:25-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, OX2RCD200 cell surface glycoprotein receptor | |
| Recombinant Mouse CD200R1 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 181 | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Cancer, GPCR, Neuronal Cell Markers, Neuroscience | |
| 0.2 um filtered solution in PBS | |
| 131450 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title