Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD200R1L Antibody (111), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289933MFV610
Description
CD200R1L Monoclonal antibody specifically detects CD200R1L in Human samples. It is validated for ELISASpecifications
CD200R1L | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
344807 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
111 | |
ELISA | |
cell surface glycoprotein CD200 receptor 2, CD200 receptor 1-like, CD200R2, CD200RLa | |
Rabbit immunized with purified, recombinant Human CD200R1L (Accession#: AAT00538.1, Met1-Leu239) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction