Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD200R1L Antibody (2692A), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB10198MFV610
Description
CD200R1L Monoclonal antibody specifically detects CD200R1L in Mouse samples. It is validated for Flow Cytometry, ImmunohistochemistrySpecifications
CD200R1L | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
0.1 mL | |
Primary | |
Mouse | |
IgG |
Flow Cytometry, Immunohistochemistry | |
2692A | |
Flow Cytometry, Immunohistochemistry | |
cell surface glycoprotein CD200 receptor 2, CD200 receptor 1-like, CD200R2, CD200RLa | |
Mouse myeloma cell line NS0-derived mouse CD200R1L, Ile25-Thr220, Accession # Q6XJV6.1 | |
RUO | |
344807 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction