Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD200R1L Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306398V
This item is not returnable.
View return policy
Description
CD200R1L Polyclonal antibody specifically detects CD200R1L in Human samples. It is validated for ELISA, ImmunofluorescenceSpecifications
CD200R1L | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
344807 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunofluorescence | |
DyLight 405 | |
cell surface glycoprotein CD200 receptor 2, CD200 receptor 1-like, CD200R2, CD200RLa | |
Produced in rabbits immunized with purified, recombinant Human CD200R1L (AAT00538.1; Met1-Leu239). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction